missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl Chloride 95.0+%, TCI America™

Catalog No. p-7124799
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C111425G 25 g
C1114100G 100 g
C1114500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C111425G Supplier TCI America Supplier No. C111425G
Only null left

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000862
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 111°C
Quantity 25 g
UN Number 2920
Formula Weight 126.96
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloropropionyl Chloride
EINECSNumber (2)-3346
TSCA Yes