missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl chloride, 95%

Catalog No. p-91370
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295161000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC295161000 Supplier Thermo Scientific Chemicals Supplier No. 295161000

CAS: 7623-09-8 | C3H4Cl2O | 126.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.97
MDL Number MFCD00000862
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C(=O)Cl)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -71.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 109.0°C to 111.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4390 to 1.4410
Linear Formula CH3CH(Cl)COCl
Quantity 100 mL
Beilstein 02, 248
Specific Gravity 1.3
Solubility Information Solubility in water: reacts
Formula Weight 126.97
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloropropionyl chloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or ha
EINECSNumber 231-540-7
TSCA TSCA

RUO – Research Use Only