missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropropene, 99%
Description
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents.
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -138°C |
| Density | 0.899 |
| Boiling Point | 22°C to 23°C |
| Flash Point | −34°C (−29°F) |
| Odor | Recognizable |
| Refractive Index | 1.3939 |
| Quantity | 5 g |
| UN Number | UN2456 |
| Beilstein | 1361376 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Show More |
Safety and Handling
| Hazard Category | H224-H319-H335 |
| Hazard Statement | GHS H Statement H224-H319-H335 Extremely flammable liquid and vapor. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 2-CHLOROPROPENE |
| EINECSNumber | 209-187-5 |
| RTECSNumber | UC7200000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only