missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropane 99.0+%, TCI America™

Catalog No. C026725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C026725ML 25 mL
C0267500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C026725ML Supplier TCI America Supplier No. C026725ML
Only null left

Chemical Identifiers

CAS 75-29-6
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.539
MDL Number MFCD00000867
InChI Key ULYZAYCEDJDHCC-UHFFFAOYSA-N
Synonym isopropyl chloride, propane, 2-chloro, isopropylchloride, 2-propyl chloride, sec-propyl chloride, chlorodimethylmethane, narcosop, isoprid, unii-vu39j8aj2n, ccris 874
PubChem CID 6361
IUPAC Name 2-chloropropane
SMILES CC(C)Cl

Specifications

Melting Point -117°C
Color Colorless
Boiling Point 35°C
Quantity 25 mL
UN Number 2356
Formula Weight 78.54
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloropropane
EINECSNumber (2)-0080
RTECSNumber TX4410000
TSCA Yes