Learn More
(±)-2-chlorophenylglycine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 433341000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| (±)-2-chlorophenylglycine | |
| 97.5 | |
| Authentic | |
| C8H8ClNO2 | |
| 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
| C1=CC=C(C(=C1)C(C(=O)O)N)Cl | |
| 185.61 | |
| 185.61 |
| 88744-36-9 | |
| 100.0 | |
| 97.5% min. (HPLC) | |
| 100g | |
| LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
| 2-amino-2-(2-chlorophenyl)acetic acid | |
| 290729 | |
| 98% |
Chemical Identifiers
| 88744-36-9 | |
| 185.61 | |
| 2-amino-2-2-chlorophenyl acetic acid, 2-2-chlorophenyl glycine, 2-chlorophenylglycine, dl-2-2-chlorophenyl glycine, amino 2-chlorophenyl acetic acid, amino-2-chloro-phenyl-acetic acid, dl-2-chlorophenylglycine, 2-chlorophenyl glycine, 2-chloro-dl-phenylglycine, h-dl-phg 2-cl-oh | |
| 2-amino-2-(2-chlorophenyl)acetic acid |
| C8H8ClNO2 | |
| LMIZLNPFTRQPSF-UHFFFAOYSA-N | |
| 290729 | |
| C1=CC=C(C(=C1)C(C(=O)O)N)Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only