missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$37.29 - $37.29
Specifications
| CAS | 3900-89-8 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C6H6BClO2 |
| MDL Number | MFCD00674012 |
| InChI Key | RRCMGJCFMJBHQC-UHFFFAOYSA-N |
Specifications
| 3900-89-8 | |
| C6H6BClO2 | |
| RRCMGJCFMJBHQC-UHFFFAOYSA-N | |
| 2734322 |
| Crystal-Powder at 20°C | |
| MFCD00674012 | |
| (2-chlorophenyl)boronic acid | |
| 156.37 |