missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenylacetic Acid 98.0+%, TCI America™
$75.08 - $292.62
Chemical Identifiers
| CAS | 2444-36-2 |
|---|---|
| Molecular Formula | C8H7ClO2 |
| Molecular Weight (g/mol) | 170.592 |
| MDL Number | MFCD00004317 |
| InChI Key | IUJAAIZKRJJZGQ-UHFFFAOYSA-N |
| Synonym | 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid |
| PubChem CID | 17124 |
| IUPAC Name | 2-(2-chlorophenyl)acetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)O)Cl |
Chemical Identifiers
| 2444-36-2 | |
| 170.592 | |
| IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
| 17124 | |
| C1=CC=C(C(=C1)CC(=O)O)Cl |
| C8H7ClO2 | |
| MFCD00004317 | |
| 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
| 2-(2-chlorophenyl)acetic acid |
Specifications
| 95°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00004317 | |
| IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
| 2-(2-chlorophenyl)acetic acid | |
| 17124 | |
| ≥98.0% (T) |
| 2444-36-2 | |
| 25 g | |
| C8H7ClO2 | |
| 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
| C1=CC=C(C(=C1)CC(=O)O)Cl | |
| 170.592 | |
| 170.59 | |
| 2-Chlorophenylacetic Acid |
Safety and Handling
EINECSNumber : (3)-1836
TSCA : No