missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. C091625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091625G 25 g
C0916500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C091625G Supplier TCI America Supplier No. C091625G

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00004317
InChI Key IUJAAIZKRJJZGQ-UHFFFAOYSA-N
Synonym 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
PubChem CID 17124
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Cl

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 170.59
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Chlorophenylacetic Acid
EINECSNumber (3)-1836
TSCA No