Learn More
2-Chlorophenylacetic acid, 98%
CAS: 2444-36-2 | C8H7ClO2 | 170.592 g/mol
$190.66 - $785.56
Chemical Identifiers
| CAS | 2444-36-2 |
|---|---|
| Molecular Formula | C8H7ClO2 |
| Molecular Weight (g/mol) | 170.592 |
| MDL Number | MFCD00004317 |
| InChI Key | IUJAAIZKRJJZGQ-UHFFFAOYSA-N |
| Synonym | 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid |
| PubChem CID | 17124 |
| IUPAC Name | 2-(2-chlorophenyl)acetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1583418
|
Thermo Scientific Chemicals
A1583418 |
50 g |
Each for $190.66
|
|
|||||
|
AAA1583430
|
Thermo Scientific Chemicals
A1583430 |
250 g |
Each for $785.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2444-36-2 | |
| 170.592 | |
| IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
| 17124 | |
| C1=CC=C(C(=C1)CC(=O)O)Cl |
| C8H7ClO2 | |
| MFCD00004317 | |
| 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
| 2-(2-chlorophenyl)acetic acid |
Specifications
| 94°C to 97°C | |
| 145°C (5 mmHg) | |
| Odorless | |
| MFCD00004317 | |
| 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid | |
| C1=CC=C(C(=C1)CC(=O)O)Cl | |
| 170.592 | |
| 170.6 | |
| 2-Chlorophenylacetic acid |
| 2444-36-2 | |
| 50 g | |
| C8H7ClO2 | |
| 1101523 | |
| IUJAAIZKRJJZGQ-UHFFFAOYSA-N | |
| 2-(2-chlorophenyl)acetic acid | |
| 17124 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-482-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only