missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic acid, 98.5+%

CAS: 2444-36-2 | C8H7ClO2 | 170.6 g/mol

$37.36 - $37.36

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00004317
InChI Key IUJAAIZKRJJZGQ-UHFFFAOYSA-N
Synonym 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
PubChem CID 17124
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC184030050
SDS View Documents
Thermo Scientific Chemicals
184030050
5 g Glass bottle
Each for $37.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2444-36-2
170.6
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
17124
C1=CC=C(C(=C1)CC(=O)O)Cl
C8H7ClO2
MFCD00004317
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
2-(2-chlorophenyl)acetic acid

Specifications

94.0°C to 97.0°C
White to Yellow
100.0
Authentic
Powder
C8H7ClO2
MFCD00004317
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
2-(2-chlorophenyl)acetic acid
17124
98.5+%
2444-36-2
98.5
5 g
Glass bottle
98.5+%
ClC6H4CH2CO2H
09, 447
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
C1=CC=C(C(=C1)CC(=O)O)Cl
170.6
170.6
2-Chlorophenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 219-482-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only