missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic acid, 98.5+%

Catalog No. AC184030050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC184030050 Supplier Thermo Scientific Chemicals Supplier No. 184030050

CAS: 2444-36-2 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
MDL Number MFCD00004317
InChI Key IUJAAIZKRJJZGQ-UHFFFAOYSA-N
Synonym 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
PubChem CID 17124
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Cl

Specifications

Melting Point 94.0°C to 97.0°C
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 98.5+%
Linear Formula ClC6H4CH2CO2H
Beilstein 09, 447
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 170.6
Percent Purity 98.5+%
Chemical Name or Material 2-Chlorophenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-482-

RUO – Research Use Only