missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl cyclopentyl ketone, 97%

Catalog No. AAL0644809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0644809 10 g
AAL0644818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0644809 Supplier Thermo Scientific Chemicals Supplier No. L0644809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 6740-85-8 | C12H13ClO | 208.685 g/mol

2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorophenyl cyclopentyl ketone is used as a reagent in the preparation of bicyclic and polycyclic aromatic hydrocarbons. It acts as a pharmaceutical intermediate and also as an intermediate of ketamine.

Solubility
Slightly miscible with chloroform and ethyl acetate.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6740-85-8
Molecular Formula C12H13ClO
Molecular Weight (g/mol) 208.685
MDL Number MFCD00038367
InChI Key QIJMMRNZBJHXRI-UHFFFAOYSA-N
Synonym 2-chlorophenyl cyclopentyl ketone, 2-chlorophenyl cyclopentyl methanone, o-chlorophenyl cyclopentyl ketone, o-chlorobenzoylcyclopentane, 2-chlorophenyl-cyclopentylketone, methanone, 2-chlorophenyl cyclopentyl, 2-chlorobenzoylcyclopentane, acmc-1b5a4, 0-chlorphenylcyclopentylketone, 1-2-chlorobenzoyl cyclopentane
PubChem CID 81223
IUPAC Name (2-chlorophenyl)-cyclopentylmethanone
SMILES C1CCC(C1)C(=O)C2=CC=CC=C2Cl

Specifications

Density 1.16
Boiling Point 130°C (5 mmHg)
Flash Point >100°C (212°F)
Odor Aromatic
Refractive Index 1.547
Quantity 10 g
Beilstein 1873022
Solubility Information Slightly miscible with chloroform and ethyl acetate.
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 2-Chlorophenyl cyclopentyl ketone
Show More Show Less
EINECSNumber 229-802-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only