missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenetole 99.0+%, TCI America™
$150.16 - $300.33
Chemical Identifiers
| CAS | 614-72-2 |
|---|---|
| Molecular Formula | C8H9ClO |
| Molecular Weight (g/mol) | 156.609 |
| MDL Number | MFCD00058930 |
| InChI Key | IRYSAAMKXPLGAM-UHFFFAOYSA-N |
| Synonym | 1-Chloro-2-ethoxybenzene |
| PubChem CID | 69197 |
| IUPAC Name | 1-chloro-2-ethoxybenzene |
| SMILES | CCOC1=CC=CC=C1Cl |
Chemical Identifiers
| 614-72-2 | |
| 156.609 | |
| IRYSAAMKXPLGAM-UHFFFAOYSA-N | |
| 69197 | |
| CCOC1=CC=CC=C1Cl |
| C8H9ClO | |
| MFCD00058930 | |
| 1-Chloro-2-ethoxybenzene | |
| 1-chloro-2-ethoxybenzene |
Specifications
| 614-72-2 | |
| 210°C | |
| Clear Liquid at 20°C | |
| MFCD00058930 | |
| IRYSAAMKXPLGAM-UHFFFAOYSA-N | |
| 1-chloro-2-ethoxybenzene | |
| 69197 | |
| ≥99.0% (GC) |
| Yellow | |
| 10 mL | |
| C8H9ClO | |
| 1-Chloro-2-ethoxybenzene | |
| CCOC1=CC=CC=C1Cl | |
| 156.609 | |
| 156.61 | |
| 2-Chlorophenetole |
Safety and Handling
EINECSNumber : (3)-0845
TSCA : No