missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenethyl bromide, 95%

Catalog No. AC440140010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440140010 1 g
AC440140050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440140010 Supplier Thermo Scientific Chemicals Supplier No. 440140010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16793-91-2 | C8H8BrCl | 219.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16793-91-2
Molecular Formula C8H8BrCl
Molecular Weight (g/mol) 219.51
InChI Key AECBVDLERUETKG-UHFFFAOYSA-N
PubChem CID 2756963
IUPAC Name 1-(2-bromoethyl)-2-chlorobenzene
SMILES C1=CC=C(C(=C1)CCBr)Cl

Specifications

Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Density 1.4569g/mL
Quantity 1 g
Boiling Point 110°C to 112°C (10.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.4569
Assay Percent Range 94% min. (GC)
Synonym 2-chlorophenethyl bromide, 1-2-bromoethyl-2-chlorobenzene, 2-2-chlorophenyl ethylbromide, 1-2-bromo-ethyl-2-chloro-benzene, 1-2-bromo-ethyl-2-chlorobenzene, benzene,1-2-bromoethyl-2-chloro, benzene, 1-2-bromoethyl-2-chloro, 1-2-bromoethyl-2-chloro-benzene, 2-chlorophenethylbromide, acmc-1cfee
Formula Weight 219.51
Percent Purity 95%
Chemical Name or Material 2-Chlorophenethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only