missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethylquinoline Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124745
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C102025G 25 g
C1020250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C102025G Supplier TCI America Supplier No. C102025G
Only null left

Chemical Identifiers

CAS 3747-74-8
Molecular Formula C10H9Cl2N
Molecular Weight (g/mol) 214.089
MDL Number MFCD00012734
InChI Key WDETYCRYUBGKCE-UHFFFAOYSA-N
Synonym 2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
PubChem CID 3083823
IUPAC Name 2-(chloromethyl)quinoline;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl

Specifications

Melting Point 193°C
Quantity 25 g
Formula Weight 214.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloromethylquinoline Hydrochloride