missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)quinoline hydrochloride, 97%

Catalog No. AAA1410214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1410214 25 g
AAA1410203 1 g
AAA1410206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1410214 Supplier Thermo Scientific Chemicals Supplier No. A1410214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3747-74-8 | C10H9Cl2N | 214.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3747-74-8
Molecular Formula C10H9Cl2N
Molecular Weight (g/mol) 214.089
MDL Number MFCD00012734
InChI Key WDETYCRYUBGKCE-UHFFFAOYSA-N
Synonym 2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
PubChem CID 3083823
IUPAC Name 2-(chloromethyl)quinoline;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl

Specifications

Melting Point 184°C to 190°C
Quantity 25 g
Beilstein 3565832
Formula Weight 214.1
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)quinoline hydrochloride
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 223-145-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only