missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)quinoline hydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. AC109510050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC109510050 Supplier Thermo Scientific Chemicals Supplier No. 109510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3747-74-8 | C10H8ClN·HCl | 214.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3747-74-8
Molecular Formula C10H8ClN·HCl
Molecular Weight (g/mol) 214.1
MDL Number MFCD00012734
InChI Key WDETYCRYUBGKCE-UHFFFAOYSA-N
Synonym 2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
PubChem CID 3083823
IUPAC Name 2-(chloromethyl)quinoline;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 183.0°C to 187.0°C
Color Cream to Yellow or Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information (5% in water) Clear
Formula Weight 214.1
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-(Chloromethyl)quinoline hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-145-3
TSCA TSCA

RUO – Research Use Only