missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)pyridine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7124629
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082225G 25 g
C0822250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082225G Supplier TCI America Supplier No. C082225G
Only null left

Chemical Identifiers

CAS 6959-47-3
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012811
InChI Key JPMRGPPMXHGKRO-UHFFFAOYSA-N
Synonym 2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
PubChem CID 23392
ChEBI CHEBI:76600
IUPAC Name 2-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC=NC(=C1)CCl.Cl

Specifications

Melting Point 125°C
Quantity 25 g
UN Number 1759
Formula Weight 164.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Chloromethyl)pyridine Hydrochloride
RTECSNumber US6825000
TSCA No