missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)benzothiazole, 95%

Catalog No. p-7056041
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6478103 1 g
AAH6478106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6478103 Supplier Thermo Scientific Chemicals Supplier No. H6478103

CAS: 37859-43-1 | C8H6ClNS | 183.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37859-43-1
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.65
MDL Number MFCD05664944
InChI Key SERUZNHRWBXDOX-UHFFFAOYSA-N
Synonym 2-chloromethyl-1,3-benzothiazole, 2-chloromethyl benzothiazole, 2-chloromethyl benzo d thiazole, benzothiazole, 2-chloromethyl, 2-chloromethylbenzothiazole, 2-chloromethyl-benzothiazole, 2-chloromethylbenzthiazole, acmc-1ag8v, ksc776s1l, 2-chloromethylbenzo d thiazole
PubChem CID 304978
IUPAC Name 2-(chloromethyl)-1,3-benzothiazole
SMILES ClCC1=NC2=CC=CC=C2S1

Specifications

Melting Point 34°C
Boiling Point 88°C to 99°C (0.3 mmHg)
Quantity 1 g
Beilstein 120805
Sensitivity Moisture sensitive
Formula Weight 183.65
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(Chloromethyl)benzothiazole
Hazard Category H302-H315-H317-H318-H335
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only