missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine Hydrochloride 98.0+%, TCI America™

Catalog No. c226925g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C226925G 25 g
C22695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C226925G Supplier TCI America Supplier No. C226925G
Only null left

Chemical Identifiers

CAS 86604-75-3
Molecular Formula C9H13Cl2NO
Molecular Weight (g/mol) 222.109
MDL Number MFCD00277472
InChI Key LCJDHJOUOJSJGS-UHFFFAOYSA-N
Synonym 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxy pyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4 methoxypyridine hydrochloride, 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hcl, pyridine, 2-chloromethyl-4-methoxy-3,5-dimethyl-, hydrochloride, 2-chloromethyl-3,5-dimethyl-4-pyridinyl methyl ether hydrochloride, pubchem2236
PubChem CID 11694258
IUPAC Name 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
SMILES CC1=CN=C(C(=C1OC)C)CCl.Cl

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 25 g
Formula Weight 222.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine Hydrochloride