missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-3,5-dioxahex-1-ene 96.0+%, TCI America™

Catalog No. p-7124851
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11981G 1 g
C11985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11981G Supplier TCI America Supplier No. C11981G

Chemical Identifiers

CAS 105104-40-3
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00130115
InChI Key JKHPOPCIKQQRPU-UHFFFAOYSA-N
Synonym 3-Chloro-2-(methoxymethoxy)-1-propene
PubChem CID 10678294
IUPAC Name 3-chloro-2-(methoxymethoxy)prop-1-ene
SMILES COCOC(=C)CCl

Specifications

Color Yellow
Boiling Point 63°C
Quantity 1 g
UN Number 1993
Formula Weight 136.58
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Chloromethyl)-3,5-dioxahex-1-ene