missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromandelic acid, 98%

Catalog No. AAB2152506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2152506 5 g
AAB2152514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2152506 Supplier Thermo Scientific Chemicals Supplier No. B2152506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10421-85-9 | C8H7ClO3 | 186.59 g/mol

2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromandelic acid, is used as a pharmaceutical intermediate as well as in the medicine industry. It is used as a intermediate of clopidrogrel.

Solubility
Very soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.

Chemical Identifiers

CAS 10421-85-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00084962
InChI Key RWOLDZZTBNYTMS-UHFFFAOYNA-N
Synonym 2-chloromandelic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, dl-2-chloromandelic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-chlorophenyl glycolic acid, 2-chloromandelicacid, 2-chloro-mandelic acid, pubchem15895
PubChem CID 97720
IUPAC Name 2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=CC=C1Cl

Specifications

Melting Point 80°C to 85°C
Quantity 5 g
Beilstein 2367070
Solubility Information Very soluble in water.
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 2-Chloromandelic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-900-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only