missing translation for 'onlineSavingsMsg'
Learn More

2-chloromalonaldehyde, 95%

Catalog No. AC330120010
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330120010 1 g Glass bottle
AC330120100 10 g Glass bottle
AC330120500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC330120010 Supplier Thermo Scientific Chemicals Supplier No. 330120010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36437-19-1 | C3H3ClO2 | 106.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36437-19-1
MDL Number MFCD01320489
InChI Key KTRZQCIGJUWSGE-UHFFFAOYSA-N
Synonym 2-chloromalonaldehyde, chloromalonaldehyde, propanedial, chloro, chloromalonalde-hyde, 2-chloro-malonaldehyde, propanedial, 2-chloro, 2-chloropropane-1,3-dial, chloromalonaldehyde; chloromalondialdehyde; chloromalonic dialdehyde;2-chloromalonaldehyde
PubChem CID 147646
IUPAC Name 2-chloropropanedial
SMILES C(=O)C(C=O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 140.0°C to 145.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H3ClO2
Linear Formula ClCH(CHO)2
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,benzene,dimethylsulfoxide
Molecular Weight (g/mol) 106.5
Formula Weight 106.5
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloromalonaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapo

RUO – Research Use Only