missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinic Acid 98.0+%, TCI America™

Catalog No. C13685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13685G 5 g
C136825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13685G Supplier TCI America Supplier No. C13685G

Chemical Identifiers

CAS 6313-54-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00191402
InChI Key QXCOHSRHFCHCHN-UHFFFAOYSA-N
Synonym 2-chloroisonicotinic acid, 2-chloro-4-pyridinecarboxylic acid, 2-chloro-isonicotinic acid, 2-chloro isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro, 2-chloro-4-pyridinyl carboxylic acid, 2-chlorisonicotins, pubchem2598, 2-chloroisonicotinic acid,, 4-carboxy-2-chloropyridine
PubChem CID 236985
IUPAC Name 2-chloropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC=NC(Cl)=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 157.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroisonicotinic Acid