missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinamide 98.0+%, TCI America™

Catalog No. p-7125815
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30791G 1 g
C30795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30791G Supplier TCI America Supplier No. C30791G

Chemical Identifiers

CAS 100859-84-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00221401
InChI Key DEMJOLRJLACBRX-UHFFFAOYSA-N
Synonym 2-chloroisonicotinamide, 4-pyridinecarboxamide, 2-chloro, 2-chloro-isonicotinamide, pubchem16074, acmc-1bpcq, 2-chloro-4-pyridinecarboxamide, 4-pyridinecarboxamide,2-chloro
PubChem CID 2800022
IUPAC Name 2-chloropyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Cl

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 1 g
Formula Weight 156.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroisonicotinamide