missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorohexane 98.0+%, TCI America™
$262.83 - $846.29
Chemical Identifiers
| CAS | 638-28-8 |
|---|---|
| Molecular Formula | C6H13Cl |
| Molecular Weight (g/mol) | 120.62 |
| MDL Number | MFCD00039351 |
| InChI Key | GLCIPJOIEVLTPR-UHFFFAOYNA-N |
| Synonym | 2-Hexyl Chloride |
| PubChem CID | 12521 |
| IUPAC Name | 2-chlorohexane |
| SMILES | CCCCC(C)Cl |
Chemical Identifiers
| 638-28-8 | |
| 120.62 | |
| GLCIPJOIEVLTPR-UHFFFAOYNA-N | |
| 12521 | |
| CCCCC(C)Cl |
| C6H13Cl | |
| MFCD00039351 | |
| 2-Hexyl Chloride | |
| 2-chlorohexane |
Specifications
| 638-28-8 | |
| 122°C | |
| MFCD00039351 | |
| 1993 | |
| GLCIPJOIEVLTPR-UHFFFAOYNA-N | |
| 2-chlorohexane | |
| 12521 | |
| ≥98.0% (GC) | |
| 2-Chlorohexane |
| Colorless | |
| C6H13Cl | |
| 5 g | |
| 2-Hexyl Chloride | |
| CCCCC(C)Cl | |
| 120.62 | |
| 120.62 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator