missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylphosphonic acid, 90%, Tech.

Catalog No. AC301630050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC301630050 5 g Glass bottle
AC301630010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC301630050 Supplier Thermo Scientific Chemicals Supplier No. 301630050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16672-87-0
Molecular Formula C2H6ClO3P
Molecular Weight (g/mol) 144.49
MDL Number MFCD00002142
InChI Key UDPGUMQDCGORJQ-UHFFFAOYSA-N
Synonym ethephon, 2-chloroethyl phosphonic acid, ethrel, chlorethephon, bromeflor, camposan, 2-cepa, phosphonic acid, 2-chloroethyl, 2-chloroethanephosphonic acid, etheverse
PubChem CID 27982
ChEBI CHEBI:52741
IUPAC Name 2-chloroethylphosphonic acid
SMILES OP(O)(=O)CCCl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 74.0°C to 75.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 3787
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in benzene,toluene,propylene glycol,practically insoluble in petroleum ether,practically insoluble in petroleum ether,freely soluble in ethanol,methanol,acetone,ethy
Formula Weight 144.51
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Chloroethylphosphonic acid, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 240-718-3
RTECSNumber SZ7100000
TSCA TSCA

RUO – Research Use Only