missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124643
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
C085125G 25 g
C0851100G 100 g
C0851500G 500 g
3 options

Catalog No. C085125G

Supplier: TCI America C085125G

Only null left

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
IUPAC Name 2-chloroethan-1-aminium chloride
SMILES [Cl-].[NH3+]CCCl

Specifications

Melting Point 147°C
Quantity 25 g
Formula Weight 115.99
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethylamine Hydrochloride
RTECSNumber KR1800000
TSCA Yes