missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylamine hydrochloride, 98+%

Catalog No. AAA1445530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1445530 250 g
AAA1445518 50 g
AAA144550B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1445530 Supplier Thermo Scientific Chemicals Supplier No. A1445530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 870-24-6 | C2H7Cl2N | 115.99 g/mol

2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
IUPAC Name 2-chloroethanamine;hydrochloride
SMILES [Cl-].[NH3+]CCCl

Specifications

Melting Point 143°C to 148°C
pH 2 to 3
Odor Odorless
Quantity 250 g
UN Number UN3261
Beilstein 3590304
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 115.99
Percent Purity ≥98%
Chemical Name or Material 2-Chloroethylamine hydrochloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H341
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-793-2
RTECSNumber KR1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only