Learn More
(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.)
CAS: 999-81-5 | C5H13Cl2N | 158.066 g/mol
$51.71 - $328.34
Chemical Identifiers
| CAS | 999-81-5 |
|---|---|
| Molecular Formula | C5H13Cl2N |
| Molecular Weight (g/mol) | 158.066 |
| MDL Number | MFCD00011869 |
| InChI Key | UHZZMRAGKVHANO-UHFFFAOYSA-M |
| Synonym | chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan |
| PubChem CID | 13836 |
| IUPAC Name | 2-chloroethyl(trimethyl)azanium;chloride |
| SMILES | C[N+](C)(C)CCCl.[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1563006
|
Thermo Scientific Chemicals
A1563006 |
5 g |
Each for $51.71
|
|
|||||
|
AAA1563022
|
Thermo Scientific Chemicals
A1563022 |
100 g |
Each for $328.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 999-81-5 | |
| 158.066 | |
| UHZZMRAGKVHANO-UHFFFAOYSA-M | |
| 13836 | |
| C[N+](C)(C)CCCl.[Cl-] |
| C5H13Cl2N | |
| MFCD00011869 | |
| chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
| 2-chloroethyl(trimethyl)azanium;chloride |
Specifications
| 999-81-5 | |
| Amine-like | |
| ClCH2CH2N(CH3)3Cl | |
| 5 g | |
| 3563994 | |
| 14,2104 | |
| UHZZMRAGKVHANO-UHFFFAOYSA-M | |
| 2-chloroethyl(trimethyl)azanium;chloride | |
| 13836 | |
| 98% | |
| (dry wt.), may cont. up to 5% water |
| ∼241°C (decomposition) | |
| C5H13Cl2N | |
| MFCD00011869 | |
| UN2811 | |
| Very Hygroscopic | |
| chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
| C[N+](C)(C)CCCl.[Cl-] | |
| 158.066 | |
| 158.07 | |
| (2-Chloroethyl)trimethylammonium chloride |
Safety and Handling
GHS H Statement
H311-H302
Toxic in contact with skin.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P361-P363-P501c
H302-H311
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 213-666-4
RTECSNumber : BP5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only