missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.)

Catalog No. AAA1563022
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1563022 100 g
AAA1563006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1563022 Supplier Thermo Scientific Chemicals Supplier No. A1563022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 999-81-5 | C5H13Cl2N | 158.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCCl.[Cl-]

Specifications

Melting Point ∼241°C (decomposition)
Odor Amine-like
Linear Formula ClCH2CH2N(CH3)3Cl
Quantity 100 g
UN Number UN2811
Beilstein 3563994
Sensitivity Very Hygroscopic
Merck Index 14,2104
Formula Weight 158.07
Percent Purity 98%
Assay (dry wt.), may cont. up to 5% water
Chemical Name or Material (2-Chloroethyl)trimethylammonium chloride
Show More Show Less
Hazard Category H302-H311
Hazard Statement GHS H Statement
H311-H302
Toxic in contact with skin.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 213-666-4
RTECSNumber BP5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only