missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)phosphonic Acid 98.0+%, TCI America™

Catalog No. p-7124995
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14561G 1 g
C14565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14561G Supplier TCI America Supplier No. C14561G

Chemical Identifiers

CAS 16672-87-0
Molecular Formula C2H6ClO3P
Molecular Weight (g/mol) 144.49
MDL Number MFCD00002142
InChI Key UDPGUMQDCGORJQ-UHFFFAOYSA-N
Synonym ethephon, 2-chloroethyl phosphonic acid, ethrel, chlorethephon, bromeflor, camposan, 2-cepa, phosphonic acid, 2-chloroethyl, 2-chloroethanephosphonic acid, etheverse
PubChem CID 27982
ChEBI CHEBI:52741
IUPAC Name (2-chloroethyl)phosphonic acid
SMILES OP(O)(=O)CCCl

Specifications

Melting Point 76°C
Color White
Boiling Point 265°C
Quantity 1 g
UN Number 2923
Formula Weight 144.49
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Chloroethyl)phosphonic Acid
EINECSNumber (2)-2930
RTECSNumber SZ7100000
TSCA No
Recommended Storage Refrigerator