missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. C238025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C238025G 25 g
C2380100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C238025G Supplier TCI America Supplier No. C238025G

Chemical Identifiers

CAS 80-41-1
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD00000970
InChI Key ZXNMIUJDTOMBPV-UHFFFAOYSA-N
Synonym 2-chloroethyl p-toluenesulfonate, 2-chloroethyl tosylate, 2-chloroethyl p-tosylate, 2-tosyloxyethyl chloride, 2-chloroethanol p-toluenesulfonate, p-toluenesulfonic acid, 2-chloroethyl ester, ethanol, 2-chloro-, 4-methylbenzenesulfonate, 2-p-toluenesulfonyloxy ethyl chloride, 2-chloroethyl-p-toluenesulfonate, p-toluenesulfonic acid-2-chloroethyl ester
PubChem CID 6639
IUPAC Name 2-chloroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCCl

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 g
Formula Weight 234.69
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl p-Toluenesulfonate
RTECSNumber XT6475000
TSCA Yes