missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl p-toluenesulfonate, 97%

Catalog No. p-7081700
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAA1186314 25 g
AAA1186322 100 g
AAA1186336 500 g
3 options

Catalog No. AAA1186314

Supplier: Thermo Scientific Chemicals A1186314

Only null left

CAS: 80-41-1 | C9H11ClO3S | 234.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-41-1
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD00000970
InChI Key ZXNMIUJDTOMBPV-UHFFFAOYSA-N
Synonym 2-chloroethyl p-toluenesulfonate, 2-chloroethyl tosylate, 2-chloroethyl p-tosylate, 2-tosyloxyethyl chloride, 2-chloroethanol p-toluenesulfonate, p-toluenesulfonic acid, 2-chloroethyl ester, ethanol, 2-chloro-, 4-methylbenzenesulfonate, 2-p-toluenesulfonyloxy ethyl chloride, 2-chloroethyl-p-toluenesulfonate, p-toluenesulfonic acid-2-chloroethyl ester
PubChem CID 6639
IUPAC Name 2-chloroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCCl

Specifications

Density 1.294
Boiling Point 158°C to 159°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 25 g
Beilstein 1968816
Formula Weight 234.7
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl p-toluenesulfonate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-277-2
RTECSNumber XT6475000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only