missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Methyl Ether 98.0+%, TCI America™

Catalog No. p-7124837
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117325ML 25 mL
C1173100ML 100 mL
C1173500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C117325ML Supplier TCI America Supplier No. C117325ML
Only null left

Chemical Identifiers

CAS 627-42-9
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000972
InChI Key XTIGGAHUZJWQMD-UHFFFAOYSA-N
Synonym 2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
PubChem CID 12316
IUPAC Name 1-chloro-2-methoxyethane
SMILES COCCCl

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 mL
UN Number 3271
Formula Weight 94.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Methyl Ether