missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%

Catalog No. p-7027376
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2408114 25 g
AAB2408122 100 g
AAB2408136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2408114 Supplier Thermo Scientific Chemicals Supplier No. B2408114

CAS: 627-42-9 | C3H7ClO | 94.54 g/mol

2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

Solubility
Soluble in water 60 g/L at 20°C.

Notes
Store away from oxidizing agents, bases.

Chemical Identifiers

CAS 627-42-9
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000972
InChI Key XTIGGAHUZJWQMD-UHFFFAOYSA-N
Synonym 2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
PubChem CID 12316
IUPAC Name 1-chloro-2-methoxyethane
SMILES COCCCl

Specifications

Density 1.04
Boiling Point 89°C to 90°C
Flash Point 8°C (46°F)
Refractive Index 1.409
Linear Formula CH3OCH2CH2Cl
Quantity 25 g
UN Number UN3271
Solubility Information Soluble in water 60g/L at 20°C.
Formula Weight 94.54
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl methyl ether
Show More Show Less
Hazard Category H225-H302
Hazard Statement GHS H Statement
H225-H302-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P312-P330-P363-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 210-999-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only