missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%, Thermo Scientific™

Chemical Identifiers

CAS 627-42-9
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000972
InChI Key XTIGGAHUZJWQMD-UHFFFAOYSA-N
Synonym 2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
PubChem CID 12316
IUPAC Name 1-chloro-2-methoxyethane
SMILES COCCCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC221771000
SDS View Documents
Thermo Scientific Chemicals
221771000
100mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC221775000
SDS View Documents
Thermo Scientific Chemicals
221775000
500mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC221770025
SDS View Documents
Thermo Scientific Chemicals
221770025
2.5L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

627-42-9
94.54
XTIGGAHUZJWQMD-UHFFFAOYSA-N
12316
COCCCl
C3H7ClO
MFCD00000972
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
1-chloro-2-methoxyethane

Specifications

627-42-9
100.0
89°C to 90°C
Authentic
Glass bottle
1.4080 to 1.41
MFCD00000972
01,337
2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
XTIGGAHUZJWQMD-UHFFFAOYSA-N
1-chloro-2-methoxyethane
12316
94.54
2-Chloroethyl methyl ether, 98%
97.5
1.0300g/mL
15°C
97.5% min. (GC)
C3H7ClO
ClCH2CH2OCH3
100mL
1.03
Solubility in water: 60g/L (20 c)
COCCCl
94.54
0.65 mPa.s (20°C)
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-999-7

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only