missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Isocyanate 98.0+%, TCI America™

$50.33 - $193.86

Chemical Identifiers

CAS 1943-83-5
Molecular Formula C3H4ClNO
Molecular Weight (g/mol) 105.521
MDL Number MFCD00002043
InChI Key BCMYXYHEMGPZJN-UHFFFAOYSA-N
Synonym 2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
PubChem CID 16035
IUPAC Name 1-chloro-2-isocyanatoethane
SMILES C(CCl)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I02935G
View Documents
TCI America
I02935G
5 g
Each for $50.33
Only null left
 
I029325G
View Documents
TCI America
I029325G
25 g
Each for $193.86
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1943-83-5
105.521
BCMYXYHEMGPZJN-UHFFFAOYSA-N
16035
C(CCl)N=C=O
C3H4ClNO
MFCD00002043
2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
1-chloro-2-isocyanatoethane

Specifications

1943-83-5
135°C
MFCD00002043
2478
BCMYXYHEMGPZJN-UHFFFAOYSA-N
1-chloro-2-isocyanatoethane
16035
≥98.0% (GC)
2-Chloroethyl Isocyanate
Colorless
C3H4ClNO
5 g
2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
C(CCl)N=C=O
105.521
105.52
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NQ8450000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'