missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl isocyanate, 97%

Catalog No. AAA1728922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1728922 100 g
AAA1728914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1728922 Supplier Thermo Scientific Chemicals Supplier No. A1728922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1943-83-5 | C3H4ClNO | 105.521 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Hydrolyzes in water.

Notes
Store at 4°C. Incompatible with alcohols. Amines, oxidizing agents. Hygroscopic.

Chemical Identifiers

CAS 1943-83-5
Molecular Formula C3H4ClNO
Molecular Weight (g/mol) 105.521
MDL Number MFCD00002043
InChI Key BCMYXYHEMGPZJN-UHFFFAOYSA-N
Synonym 2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
PubChem CID 16035
IUPAC Name 1-chloro-2-isocyanatoethane
SMILES C(CCl)N=C=O

Specifications

Density 1.237
Boiling Point 135°C
Flash Point 56°C (132°F)
Odor Pungent
Refractive Index 1.447
Quantity 100 g
UN Number UN3080
Beilstein 1071429
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 105.52
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl isocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H314-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H314-H318-H226
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 217-734-4
RTECSNumber NQ8450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only