missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethyl Chloromethyl Ether 98.0+%, TCI America™
$126.76 - $363.49
Chemical Identifiers
| CAS | 1462-33-5 |
|---|---|
| Molecular Formula | C3H6Cl2O |
| Molecular Weight (g/mol) | 128.98 |
| MDL Number | MFCD00191406 |
| InChI Key | LUTWEKBTDWRTSE-UHFFFAOYSA-N |
| Synonym | 2-(Chloromethoxy)ethyl Chloride |
| PubChem CID | 10290810 |
| IUPAC Name | 1-chloro-2-(chloromethoxy)ethane |
| SMILES | ClCCOCCl |
Chemical Identifiers
| 1462-33-5 | |
| 128.98 | |
| LUTWEKBTDWRTSE-UHFFFAOYSA-N | |
| 10290810 | |
| ClCCOCCl |
| C3H6Cl2O | |
| MFCD00191406 | |
| 2-(Chloromethoxy)ethyl Chloride | |
| 1-chloro-2-(chloromethoxy)ethane |
Specifications
| 1462-33-5 | |
| 147°C | |
| MFCD00191406 | |
| 2929 | |
| LUTWEKBTDWRTSE-UHFFFAOYSA-N | |
| 1-chloro-2-(chloromethoxy)ethane | |
| 10290810 | |
| ≥98.0% (GC) | |
| 2-Chloroethyl Chloromethyl Ether |
| Colorless | |
| C3H6Cl2O | |
| 5 g | |
| 2-(Chloromethoxy)ethyl Chloride | |
| ClCCOCCl | |
| 128.98 | |
| 128.98 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator