missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)benzene 99.0+%, TCI America™

Catalog No. C016725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C016725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C016725G Supplier TCI America Supplier No. C016725G
Only null left

Chemical Identifiers

CAS 622-24-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000977
InChI Key MNNZINNZIQVULG-UHFFFAOYSA-N
Synonym 2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
PubChem CID 231496
IUPAC Name (2-chloroethyl)benzene
SMILES ClCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 140.61
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Chloroethyl)benzene
EINECSNumber (3)-0033
TSCA Yes