missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloroethyl)benzene, 99%

Catalog No. AAL0489136
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0489136 500 g
AAL0489109 10 g
AAL0489122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0489136 Supplier Thermo Scientific Chemicals Supplier No. L0489136
Only null left

CAS: 622-24-2 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-24-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000977
InChI Key MNNZINNZIQVULG-UHFFFAOYSA-N
Synonym 2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
PubChem CID 231496
IUPAC Name 2-chloroethylbenzene
SMILES ClCCC1=CC=CC=C1

Specifications

Melting Point -60°C
Density 1.07
Boiling Point 82°C to 84°C (16 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.53
Quantity 500 g
Beilstein 385969
Formula Weight 140.61
Percent Purity 99%
Chemical Name or Material 2-(Chloroethyl)benzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-725-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only