missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanesulfonyl Chloride 95.0+%, TCI America™

Catalog No. C114225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C114225G 25 g
C1142100G 100 g
C1142500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C114225G Supplier TCI America Supplier No. C114225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1622-32-8
Molecular Formula C2H4Cl2O2S
Molecular Weight (g/mol) 163.012
MDL Number MFCD00007461
InChI Key VHCSBTPOPKFYIU-UHFFFAOYSA-N
Synonym 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
PubChem CID 15385
IUPAC Name 2-chloroethanesulfonyl chloride
SMILES C(CCl)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 203°C
Quantity 25 g
UN Number 2927
Formula Weight 163.01
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloroethanesulfonyl Chloride
RTECSNumber KI8060000
TSCA Yes
Recommended Storage Refrigerator