Learn More
2-Chloroethanesulfonyl chloride, 98%
CAS: 1622-32-8 | C2H4Cl2O2S | 163.012 g/mol
Supplier: Thermo Scientific Chemicals A1537722
Description
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
Specifications
| 2-Chloroethanesulfonyl chloride | |
| 1.56 | |
| >110°C (230°F) | |
| 1.4915 | |
| MFCD00007461 | |
| UN3390 | |
| Moisture sensitive | |
| Reacts with water. | |
| C(CCl)S(=O)(=O)Cl | |
| 163.012 | |
| 163.02 |
| 1622-32-8 | |
| 203°C | |
| C2H4Cl2O2S | |
| ClCH2CH2SO2Cl | |
| 100 g | |
| 1751202 | |
| 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride | |
| VHCSBTPOPKFYIU-UHFFFAOYSA-N | |
| 2-chloroethanesulfonyl chloride | |
| 15385 | |
| 98% |
Chemical Identifiers
| 1622-32-8 | |
| 163.012 | |
| VHCSBTPOPKFYIU-UHFFFAOYSA-N | |
| 15385 | |
| C(CCl)S(=O)(=O)Cl |
| C2H4Cl2O2S | |
| MFCD00007461 | |
| 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride | |
| 2-chloroethanesulfonyl chloride |
Safety and Handling
GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H330-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 216-594-1
RTECSNumber : KI8060000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only