missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclopentanone (stabilized with HQ + CaCO3) 95.0+%, TCI America™

Catalog No. p-7124980
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14375G 5 g
C143725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14375G Supplier TCI America Supplier No. C14375G
Only null left

Chemical Identifiers

CAS 694-28-0
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001410
InChI Key AXDZFGRFZOQVBV-UHFFFAOYNA-N
Synonym 2-chlorocyclopentanone, cyclopentanone, 2-chloro, 2-chloro-1-cyclopentanone, alpha-chlorocyclopentanone, 2-chlorocyclopentan-one, .alpha.-chlorocyclopentanone, chlorocyclopentanone, o-chlorocyclopentanone, wln: l5vtj bg, acmc-209u9b
PubChem CID 12751
IUPAC Name 2-chlorocyclopentan-1-one
SMILES ClC1CCCC1=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 118.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclopentanone (stabilized with HQ + CaCO3)
RTECSNumber GY4900000
TSCA Yes
Recommended Storage Freezer