missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclopentanone, 97%, stabilized

Catalog No. p-86629
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108970050 5 g Glass bottle
AC108970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108970050 Supplier Thermo Scientific Chemicals Supplier No. 108970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 694-28-0 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 694-28-0
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001410
InChI Key AXDZFGRFZOQVBV-UHFFFAOYNA-N
Synonym 2-chlorocyclopentanone, cyclopentanone, 2-chloro, 2-chloro-1-cyclopentanone, alpha-chlorocyclopentanone, 2-chlorocyclopentan-one, .alpha.-chlorocyclopentanone, chlorocyclopentanone, o-chlorocyclopentanone, wln: l5vtj bg, acmc-209u9b
PubChem CID 12751
IUPAC Name 2-chlorocyclopentan-1-one
SMILES ClC1CCCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.1800g/mL
Boiling Point 72.0°C to 74.0°C (12.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.4770
Linear Formula ClC5H7(=O)
Quantity 5 g
Beilstein 07, I, 5
Specific Gravity 1.18
Stabilizer about 1% dry potassium carbonate
Formula Weight 118.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclopentanone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-769-9
RTECSNumber GY4900000
TSCA TSCA
Recommended Storage Product may discolor on storage

RUO – Research Use Only