missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone (stabilized with HQ + CaCO3) 96.0+%, TCI America™

Supplier:  TCI America C096525G

Catalog No. C096525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Chlorocyclohexanone (stabilized with HQ + CaCO3)
Yellow
C6H9ClO
25 g
CCHNWURRBFGQCD-UHFFFAOYNA-N
2-chlorocyclohexan-1-one
13203
≥96.0% (GC)
822-87-7
107°C
MFCD00001626
2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
ClC1CCCCC1=O
132.59
132.59
Liquid

Chemical Identifiers

822-87-7
132.59
CCHNWURRBFGQCD-UHFFFAOYNA-N
13203
ClC1CCCCC1=O
C6H9ClO
MFCD00001626
2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
2-chlorocyclohexan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2408

RTECSNumber : GW1225000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions