missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorocyclohexanone, 97%, stab. with calcium carbonate/magnesium oxide
Description
2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep refrigerated at -20°C.
2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep refrigerated at -20°C.
Specifications
Specifications
| Melting Point | 22°C to 24°C |
| Density | 1.161 |
| Boiling Point | 82°C to 83°C (10 mmHg) |
| Flash Point | 82°C (180°F) |
| Refractive Index | 1.484 |
| Quantity | 10 g |
| Beilstein | 774100 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 132.59 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H317-H335 Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c |
| EINECSNumber | 212-505-5 |
| RTECSNumber | GW1225000 |
| TSCA | Yes |
| Recommended Storage | Store at -20°C; Store under Argon |
RUO – Research Use Only