missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone, 97%, stab. with calcium carbonate/magnesium oxide

Catalog No. AAA1861309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1861309 10 g
AAA1861318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1861309 Supplier Thermo Scientific Chemicals Supplier No. A1861309

CAS: 822-87-7 | C6H9ClO | 132.59 g/mol

2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep refrigerated at -20°C.

Chemical Identifiers

CAS 822-87-7
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.59
MDL Number MFCD00001626
InChI Key CCHNWURRBFGQCD-UHFFFAOYNA-N
Synonym 2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
PubChem CID 13203
IUPAC Name 2-chlorocyclohexan-1-one
SMILES ClC1CCCCC1=O

Specifications

Melting Point 22°C to 24°C
Density 1.161
Boiling Point 82°C to 83°C (10 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.484
Quantity 10 g
Beilstein 774100
Solubility Information Insoluble in water.
Formula Weight 132.59
Percent Purity 97%
Chemical Name or Material 2-Chlorocyclohexanone, stabilized with calcium carbonate/magnesium oxide
Show More Show Less
Hazard Category H227-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
EINECSNumber 212-505-5
RTECSNumber GW1225000
TSCA Yes
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only