missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone, 95%, stabilized

Catalog No. AC108961000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108961000 100 g Plastic bottle
AC108960050 5 g Plastic bottle
AC108960250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108961000 Supplier Thermo Scientific Chemicals Supplier No. 108961000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 822-87-7 | C6H9ClO | 132.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 822-87-7
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.59
MDL Number MFCD00001626
InChI Key CCHNWURRBFGQCD-UHFFFAOYNA-N
Synonym 2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
PubChem CID 13203
IUPAC Name 2-chlorocyclohexan-1-one
SMILES ClC1CCCCC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 23°C
Color Brown
Density 1.1600g/mL
Boiling Point 82°C to 83°C (10.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4830 to 1.485
Linear Formula ClC6H9(=O)
Quantity 100 g
Beilstein 07, 10
Specific Gravity 1.16
Stabilizer 0.2% CaCO3/MgO (1:1)
Formula Weight 132.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclohexanone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-505-5
RTECSNumber GW1225000
TSCA TSCA

RUO – Research Use Only